Assay Details
| Gene Symbol: YIPF6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022148.4 | XP_004022197.1 | 3 - 4 | 333 | 86 | |
| XM_012106839.3 | XP_011962229.1 | 3 - 4 | 338 | 86 | ||
More Information
Set Membership: |
|

YIPF6
Genomic Map