Assay Details
| Gene Symbol: GMFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004010658.3 | XP_004010707.1 | 3 - 4 | 255 | 75 | |
| XM_012181683.3 | XP_012037073.1 | 3 - 4 | 255 | 75 | ||
More Information
Set Membership: |
|

GMFB
Genomic Map