Assay Details
| Gene Symbol: LRRC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973352.1 | XP_027829153.1 | 1 - 2 | 2298 | 66 | |
| XM_027973354.1 | XP_027829155.1 | 1 - 2 | 2298 | 66 | ||
| XM_027973355.1 | XP_027829156.1 | 1 - 2 | 2298 | 66 | ||
| XM_027973356.1 | XP_027829157.1 | 2 - 3 | 296 | 66 | ||
| XM_027973357.1 | XP_027829158.1 | 2 - 3 | 372 | 66 | ||
More Information
Set Membership: |
|

LRRC24
Genomic Map