Assay Details
Gene Symbol: LOC101118315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027959706.1 | XP_027815507.1 | 4 - 5 | 532 | 87 | |
XM_027959707.1 | XP_027815508.1 | 5 - 6 | 721 | 87 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
