Assay Details
Gene Symbol: ROMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012189126.3 | XP_012044516.1 | 3 - 4 | 670 | 88 | |
XM_012189128.3 | XP_012044518.1 | 2 - 3 | 302 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
