Assay Details
| Gene Symbol: LAGE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962774.1 | XP_027818575.1 | 1 - 2 | 300 | 65 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LAGE3| Gene Symbol: LAGE3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962774.1 | XP_027818575.1 | 1 - 2 | 300 | 65 | |