Assay Details
| Gene Symbol: EFEMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012171221.3 | XP_012026611.3 | 4 - 5 | 553 | 59 | |
| XM_027966816.1 | XP_027822617.1 | 4 - 5 | 617 | 59 | ||
| XM_027966817.1 | XP_027822618.1 | 4 - 5 | 803 | 59 | ||
| XM_027966819.1 | XP_027822620.1 | 4 - 5 | 617 | 59 | ||
More Information
Set Membership: |
|

EFEMP1
Genomic Map