Assay Details
| Gene Symbol: NGRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004017754.4 | XP_004017803.2 | 1 - 2 | 711 | 71 | |
| Gene Symbol: LOC114109001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027957058.1 | XP_027812859.1 | 1 - 2 | 212 | 71 | |

NGRN
Genomic Map