Assay Details
| Gene Symbol: FAM20A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027973995.1 | XP_027829796.1 | 4 - 5 | 732 | 56 | |
| XM_027973996.1 | XP_027829797.1 | 2 - 3 | 1403 | 56 | ||
| XM_027973997.1 | XP_027829798.1 | 4 - 5 | 456 | 56 | ||
More Information
Set Membership: |
|

FAM20A
Genomic Map