Assay Details
| Gene Symbol: SLC41A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004006688.4 | XP_004006737.1 | 5 - 6 | 1108 | 63 | |
| XM_027967679.1 | XP_027823480.1 | 5 - 6 | 1307 | 63 | ||
| XM_027967680.1 | XP_027823481.1 | 5 - 6 | 1161 | 63 | ||
More Information
Set Membership: |
|

SLC41A2
Genomic Map