Assay Details
| Gene Symbol: ZNF185 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012107127.3 | XP_011962517.2 | 13 - 14 | 1022 | 61 | |
| XM_012107129.3 | XP_011962519.2 | 13 - 14 | 1019 | 61 | ||
| XM_012107130.3 | XP_011962520.2 | 13 - 14 | 1019 | 61 | ||
| XM_012107131.3 | XP_011962521.2 | 13 - 14 | 1016 | 61 | ||
| XM_012107141.3 | XP_011962531.2 | 13 - 14 | 1022 | 61 | ||
| XM_027962922.1 | XP_027818723.1 | 12 - 13 | 947 | 61 | ||
| XM_027962923.1 | XP_027818724.1 | 14 - 15 | 1100 | 61 | ||
More Information
Set Membership: |
|

ZNF185
Genomic Map