Assay Details
| Gene Symbol: CCDC113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189846.3 | XP_012045236.2 | 3 - 4 | 465 | 57 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCDC113| Gene Symbol: CCDC113 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012189846.3 | XP_012045236.2 | 3 - 4 | 465 | 57 | |
Set Membership: |
|