Assay Details
| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977754.1 | XP_027833555.1 | 7 - 8 | 1206 | 66 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A31| Gene Symbol: SLC22A31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977754.1 | XP_027833555.1 | 7 - 8 | 1206 | 66 | |
Set Membership: |
|