Assay Details
| Gene Symbol: TCF19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018995.4 | XP_004019044.2 | 2 - 3 | 705 | 105 | |
| XM_027958906.1 | XP_027814707.1 | 2 - 3 | 558 | 105 | ||
More Information
Set Membership: |
|

TCF19
Genomic Map