Assay Details
| Gene Symbol: CCZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015103822.2 | XP_014959308.2 | 11 - 12 | 1050 | 58 | |
| XM_027961779.1 | XP_027817580.1 | 11 - 12 | 1025 | 58 | ||
More Information
Set Membership: |
|

CCZ1
Genomic Map