Assay Details
| Gene Symbol: OGFRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027972793.1 | XP_027828594.1 | 4 - 5 | 542 | 62 | |
| XM_027972794.1 | XP_027828595.1 | 4 - 5 | 539 | 62 | ||
More Information
Set Membership: |
|

OGFRL1
Genomic Map