Assay Details
| Gene Symbol: STPG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970734.1 | XP_027826535.1 | 3 - 4 | 379 | 85 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

STPG2| Gene Symbol: STPG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027970734.1 | XP_027826535.1 | 3 - 4 | 379 | 85 | |
Set Membership: |
|