Assay Details
| Gene Symbol: ZNF143 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004016165.4 | XP_004016214.1 | 14 - 15 | 1778 | 56 | |
| XM_012095789.3 | XP_011951179.1 | 14 - 15 | 1764 | 56 | ||
| XM_012095790.3 | XP_011951180.1 | 14 - 15 | 1761 | 56 | ||
| XM_012095792.3 | XP_011951182.1 | 14 - 15 | 1775 | 56 | ||
More Information
Set Membership: |
|

ZNF143
Genomic Map