Assay Details
Gene Symbol: ZNF827 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_012097410.3 | XP_011952800.3 | 9 - 10 | 3038 | 59 | |
XM_027956250.1 | XP_027812051.1 | 9 - 10 | 1880 | 59 | ||
XM_027956251.1 | XP_027812052.1 | 9 - 10 | 3038 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
