Assay Details
| Gene Symbol: MAFG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012186426.3 | XP_012041816.2 | 3 | 5782 | 77 | |
| XM_015098909.2 | XP_014954395.2 | 3 | 2745 | 77 | ||
| XM_027974284.1 | XP_027830085.1 | 2 | 2672 | 77 | ||
More Information
Set Membership: |
|

MAFG
Genomic Map