Assay Details
| Gene Symbol: KIAA0753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012185538.3 | XP_012040928.2 | 9 - 10 | 1851 | 57 | |
| XM_012185539.3 | XP_012040929.2 | 9 - 10 | 1878 | 57 | ||
| XM_027975098.1 | XP_027830899.1 | 9 - 10 | 1878 | 57 | ||
More Information
Set Membership: |
|

KIAA0753
Genomic Map