Assay Details
| Gene Symbol: PGPEP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015101866.2 | XP_014957352.2 | 3 - 4 | 500 | 62 | |
| XM_027957016.1 | XP_027812817.1 | 5 - 6 | 2036 | 62 | ||
| XM_027957017.1 | XP_027812818.1 | 5 - 6 | 1673 | 62 | ||
| XM_027957018.1 | XP_027812819.1 | 4 - 5 | 1983 | 62 | ||
| XM_027957020.1 | XP_027812821.1 | 4 - 5 | 1679 | 62 | ||
| XM_027957021.1 | XP_027812822.1 | 4 - 5 | 1620 | 62 | ||
| XM_027957022.1 | XP_027812823.1 | 3 - 4 | 521 | 62 | ||
| XM_027957023.1 | XP_027812824.1 | 2 - 3 | 458 | 62 | ||
| XM_027957024.1 | XP_027812825.1 | 5 - 6 | 2017 | 62 | ||
| XM_027957025.1 | XP_027812826.1 | 5 - 6 | 1285 | 62 | ||
| XM_027957026.1 | XP_027812827.1 | 4 - 5 | 684 | 62 | ||
| XM_027957027.1 | XP_027812828.1 | 2 - 3 | 452 | 62 | ||
| XM_027957028.1 | XP_027812829.1 | 2 - 3 | 267 | 62 | ||
| XM_027957029.1 | XP_027812830.1 | 2 - 3 | 880 | 62 | ||
| XM_027957030.1 | XP_027812831.1 | 2 - 3 | 444 | 62 | ||
More Information
Set Membership: |
|

PGPEP1L
Genomic Map