Assay Details
| Gene Symbol: TEX15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962577.1 | XP_027818378.1 | 9 - 10 | 9592 | 73 | |
| XM_027962578.1 | XP_027818379.1 | 10 - 11 | 9461 | 73 | ||
| XM_027962579.1 | XP_027818380.1 | 8 - 9 | 8843 | 73 | ||
More Information
Set Membership: |
|

TEX15
Genomic Map