Assay Details
| Gene Symbol: KLK13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977525.1 | XP_027833326.1 | 4 - 5 | 677 | 75 | |
| XM_027977526.1 | XP_027833327.1 | 2 - 3 | 200 | 75 | ||
More Information
Set Membership: |
|

KLK13
Genomic Map