Assay Details
| Gene Symbol: TTC23L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012097099.3 | XP_011952489.2 | 5 - 6 | 697 | 64 | |
| XM_027980126.1 | XP_027835927.1 | 4 - 5 | 1636 | 64 | ||
More Information
Set Membership: |
|

TTC23L
Genomic Map