Assay Details
| Gene Symbol: ZNF527 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012190329.2 | XP_012045719.2 | 5 - 6 | 891 | 68 | |
| XM_027977402.1 | XP_027833203.1 | 5 - 6 | 1294 | 68 | ||
| XM_027977403.1 | XP_027833204.1 | 3 - 4 | 716 | 68 | ||
More Information
Set Membership: |
|

ZNF527
Genomic Map