Assay Details
| Gene Symbol: ZBED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019051.4 | XP_004019100.2 | 3 - 4 | 2616 | 96 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZBED9| Gene Symbol: ZBED9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004019051.4 | XP_004019100.2 | 3 - 4 | 2616 | 96 | |