Assay Details
Gene Symbol: SEPTIN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_027974155.1 | XP_027829956.1 | 9 - 10 | 1523 | 55 | |
XM_027974156.1 | XP_027829957.1 | 8 - 9 | 1651 | 55 | ||
XM_027974157.1 | XP_027829958.1 | 8 - 9 | 1522 | 55 | ||
XM_027974158.1 | XP_027829959.1 | 8 - 9 | 1430 | 55 | ||
XM_027974159.1 | XP_027829960.1 | 7 - 8 | 921 | 55 | ||
XM_027974160.1 | XP_027829961.1 | 7 - 8 | 782 | 55 | ||
XM_027974161.1 | XP_027829962.1 | 7 - 8 | 866 | 55 | ||
XM_027974162.1 | XP_027829963.1 | 7 - 8 | 958 | 55 | ||
XM_027974163.1 | XP_027829964.1 | 7 - 8 | 1235 | 55 | ||
XM_027974164.1 | XP_027829965.1 | 7 - 8 | 805 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
