Assay Details
| Gene Symbol: ANKRD33 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012174248.2 | XP_012029638.2 | 3 - 4 | 358 | 66 | |
| XM_015094654.2 | XP_014950140.2 | 5 - 6 | 1724 | 66 | ||
| XM_027967299.1 | XP_027823100.1 | 6 - 7 | 1326 | 66 | ||
| XM_027967300.1 | XP_027823101.1 | 5 - 6 | 1668 | 66 | ||
More Information
Set Membership: |
|

ANKRD33
Genomic Map