Assay Details
| Gene Symbol: FAM221A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012176922.3 | XP_012032312.1 | 2 - 3 | 332 | 74 | |
| XM_012176928.3 | XP_012032318.1 | 2 - 3 | 296 | 74 | ||
| XM_027968674.1 | XP_027824475.1 | 2 - 3 | 296 | 74 | ||
| XM_027968675.1 | XP_027824476.1 | 2 - 3 | 296 | 74 | ||
| XM_027968676.1 | XP_027824477.1 | 2 - 3 | 264 | 74 | ||
More Information
Set Membership: |
|

FAM221A
Genomic Map