Assay Details
| Gene Symbol: LDLRAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093691.2 | XP_014949177.1 | 1 - 2 | 91 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LDLRAD2| Gene Symbol: LDLRAD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015093691.2 | XP_014949177.1 | 1 - 2 | 91 | 58 | |
Set Membership: |
|