Assay Details
| Gene Symbol: SLC22A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958019.1 | XP_027813820.1 | 2 - 3 | 626 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC22A14| Gene Symbol: SLC22A14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027958019.1 | XP_027813820.1 | 2 - 3 | 626 | 63 | |
Set Membership: |
|