Assay Details
| Gene Symbol: EXOSC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018523.4 | XP_004018572.3 | 7 - 8 | 808 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EXOSC7| Gene Symbol: EXOSC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004018523.4 | XP_004018572.3 | 7 - 8 | 808 | 69 | |
Set Membership: |
|