Assay Details
| Gene Symbol: TMEM104 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974042.1 | XP_027829843.1 | 3 - 4 | 335 | 61 | |
| XM_027974043.1 | XP_027829844.1 | 3 - 4 | 300 | 61 | ||
More Information
Set Membership: |
|

TMEM104
Genomic Map