Assay Details
| Gene Symbol: CD300A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974031.1 | XP_027829832.1 | 2 - 3 | 683 | 62 | |
| XM_027974032.1 | XP_027829833.1 | 2 - 3 | 682 | 62 | ||
More Information
Set Membership: |
|

CD300A
Genomic Map