Assay Details
| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975359.1 | XP_027831160.1 | 5 - 6 | 1541 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RNF43| Gene Symbol: RNF43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027975359.1 | XP_027831160.1 | 5 - 6 | 1541 | 67 | |
Set Membership: |
|