Assay Details
| Gene Symbol: NPY1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004009578.4 | XP_004009627.1 | 2 - 3 | 1020 | 87 | |
| XM_012179386.3 | XP_012034776.1 | 3 - 4 | 1153 | 87 | ||
| XM_012179388.3 | XP_012034778.1 | 3 - 4 | 1373 | 87 | ||
More Information
Set Membership: |
|

NPY1R
Genomic Map