Assay Details
| Gene Symbol: HSDL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977678.1 | XP_027833479.1 | 3 - 4 | 788 | 58 | |
| XM_027977679.1 | XP_027833480.1 | 4 - 5 | 792 | 58 | ||
More Information
Set Membership: |
|

HSDL1
Genomic Map