Assay Details
| Gene Symbol: FAM229B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012182573.3 | XP_012037963.2 | 3 - 4 | 427 | 101 | |
| XM_012182574.3 | XP_012037964.2 | 3 - 4 | 438 | 101 | ||
| XM_012182575.3 | XP_012037965.2 | 3 - 4 | 424 | 101 | ||
| XM_012182576.3 | XP_012037966.2 | 3 - 4 | 435 | 101 | ||
| XM_012182584.3 | XP_012037974.2 | 2 - 3 | 269 | 101 | ||
| XM_027972355.1 | XP_027828156.1 | 3 - 4 | 343 | 101 | ||
More Information
Set Membership: |
|

FAM229B
Genomic Map