Assay Details
| Gene Symbol: NDUFA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004008868.3 | XP_004008917.1 | 1 - 2 | 143 | 87 | |
| XM_012178619.3 | XP_012034009.1 | 1 - 2 | 151 | 87 | ||
More Information
Set Membership: |
|

NDUFA2
Genomic Map