Assay Details
| Gene Symbol: B3GAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027979836.1 | XP_027835637.1 | 5 - 6 | 1085 | 95 | |
| XM_027979837.1 | XP_027835638.1 | 5 - 6 | 1158 | 95 | ||
More Information
Set Membership: |
|

B3GAT1
Genomic Map