Assay Details
| Gene Symbol: CHMP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974252.1 | XP_027830053.1 | 7 - 8 | 696 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CHMP6| Gene Symbol: CHMP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027974252.1 | XP_027830053.1 | 7 - 8 | 696 | 61 | |
Set Membership: |
|