Assay Details
| Gene Symbol: TRIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969302.1 | XP_027825103.1 | 2 - 3 | 1228 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM17| Gene Symbol: TRIM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969302.1 | XP_027825103.1 | 2 - 3 | 1228 | 61 | |
Set Membership: |
|