Assay Details
| Gene Symbol: MPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027962018.1 | XP_027817819.1 | 2 - 3 | 363 | 59 | |
| XM_027962019.1 | XP_027817820.1 | 2 - 3 | 352 | 59 | ||
More Information
Set Membership: |
|

MPG
Genomic Map