Assay Details
| Gene Symbol: MRPL54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027969871.1 | XP_027825672.1 | 2 - 3 | 318 | 100 | |
| XM_027969872.1 | XP_027825673.1 | 2 - 3 | 315 | 100 | ||
More Information
Set Membership: |
|

MRPL54
Genomic Map