Assay Details
| Gene Symbol: SLC38A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977675.1 | XP_027833476.1 | 7 - 8 | 1020 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC38A8| Gene Symbol: SLC38A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_027977675.1 | XP_027833476.1 | 7 - 8 | 1020 | 60 | |
Set Membership: |
|