Assay Details
| Gene Symbol: ZSCAN16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_012101094.3 | XP_011956484.2 | 3 - 4 | 3338 | 82 | |
| XM_027959061.1 | XP_027814862.1 | 2 - 3 | 781 | 82 | ||
More Information
Set Membership: |
|

ZSCAN16
Genomic Map