Assay Details
| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014976.4 | XP_004015025.1 | 4 - 5 | 386 | 68 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CNEP1R1| Gene Symbol: CNEP1R1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004014976.4 | XP_004015025.1 | 4 - 5 | 386 | 68 | |
Set Membership: |
|