Assay Details
Gene Symbol: CWC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004015988.4 | XP_004016037.1 | 6 - 7 | 679 | 81 | |
XM_012095360.3 | XP_011950750.1 | 6 - 7 | 683 | 81 | ||
XM_012095362.2 | XP_011950752.1 | 6 - 7 | 717 | 81 | ||
XM_012095364.3 | XP_011950754.1 | 6 - 7 | 825 | 81 | ||
XM_015100886.2 | XP_014956372.1 | 6 - 7 | 1170 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
