Assay Details
| Gene Symbol: ASB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022030.4 | XP_004022079.2 | 2 - 3 | 989 | 97 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ASB12| Gene Symbol: ASB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004022030.4 | XP_004022079.2 | 2 - 3 | 989 | 97 | |
Set Membership: |
|